4-Butoxy-2,3-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37453-1G 37453-5G 37453-25G
77340-992EA 66.95 USD
77340-992 77362-698 77374-290
4-Butoxy-2,3-difluorobenzoic acid ≥97%
4-Butoxy-2,3-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34529934
CAS Number: 331245-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR