1-Butoxy-4-chloro-2,3-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

37451-500MG 37451-1G 37451-5G 37451-25G
77326-386EA 249.26 USD
77326-386 77350-080 77362-694 77385-536
1-Butoxy-4-chloro-2,3-difluorobenzene ≥97%
1-Butoxy-4-chloro-2,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClF₂O
MW: 220.644 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044161
CAS Number: 1881295-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR