1-Butoxy-2,3-difluoro-4-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

37450-1G 37450-5G 37450-25G
77340-894EA 62.83 USD
77340-894 77362-692 77374-288
1-Butoxy-2,3-difluoro-4-iodobenzene ≥95%
1-Butoxy-2,3-difluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₂IO
MW: 312.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707026
CAS Number: 1366126-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR