(5-Bromo-2,3-difluoro-4-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

37443-500MG 37443-1G 37443-5G
77326-384EA 332.69 USD
77326-384 77350-078 77374-284
(5-Bromo-2,3-difluoro-4-methoxyphenyl)methanol ≥97%
(5-Bromo-2,3-difluoro-4-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O₂
MW: 253.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632223
CAS Number: 1781546-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR