2,3-Dichloro-4-(methoxymethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

37425-500MG 37425-1G 37425-5G
77335-932EA 369.77 USD
77335-932 77350-070 77374-280
2,3-Dichloro-4-(methoxymethyl)pyridine ≥95%
2,3-Dichloro-4-(methoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MW: 192.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564049
CAS Number: 2734777-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR