Ethyl 4-bromo-2-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37418-500MG 37418-1G 37418-5G 37418-25G
77326-374EA 218.36 USD
77326-374 77350-068 77362-682 77385-528
Ethyl 4-bromo-2-isopropylbenzoate ≥97%
Ethyl 4-bromo-2-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrO₂
MW: 271.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632215
CAS Number: 2404733-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR