tert-Butyl 4-bromo-2-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37417-500MG 37417-1G 37417-5G 37417-25G
77326-372EA 202.91 USD
77326-372 77350-066 77362-680 77385-526
tert-Butyl 4-bromo-2-isopropylbenzoate ≥97%
tert-Butyl 4-bromo-2-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₉BrO₂
MW: 299.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564047
CAS Number: 2734776-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR