5-Chloro-2-isopropoxy-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

37416-500MG 37416-1G 37416-5G 37416-25G
77326-370EA 215.27 USD
77326-370 77350-064 77362-678 77385-524
5-Chloro-2-isopropoxy-4-methylpyridine ≥97%
5-Chloro-2-isopropoxy-4-methylpyridine

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO
MW: 185.651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632214
CAS Number: 2404734-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR