2-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-methyl-5-nitropyridine ≥97%

Supplier: AOBChem USA

Warning

37409-500MG 37409-1G 37409-5G 37409-25G
77326-368EA 119.48 USD
77326-368 77343-644 77362-676 77374-278
2-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-methyl-5-nitropyridine ≥97%
2-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-methyl-5-nitropyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₃N₃O₂
MW: 231.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632213
CAS Number: 2404733-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR