4-Methoxy-N,3,5-trimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

37394-500MG 37394-1G 37394-5G 37394-25G
77326-366EA 218.36 USD
77326-366 77350-062 77362-672 77385-522
4-Methoxy-N,3,5-trimethylbenzamide ≥97%
4-Methoxy-N,3,5-trimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂
MW: 193.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632210
CAS Number: 1888958-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR