(4-Methoxy-3,5-dimethylphenyl)(thiomorpholino)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
37392-500MG
37392-1G
37392-5G
37392-25G
77326-364EA
243.08
USD
77326-364
77350-060
77362-670
77385-520
(4-Methoxy-3,5-dimethylphenyl)(thiomorpholino)methanone ≥97%
(4-Methoxy-3,5-dimethylphenyl)(thiomorpholino)methanone
Precaución: for Research Use Only
Formula:
C₁₄H₁₉NO₂S MW: 265.371 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32632209 CAS Number: 2325522-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...