4-Methoxy-3,5-dimethylbenzamide ≥95%

Supplier: AOBChem USA

Warning

37391-500MG 37391-1G 37391-5G 37391-25G
77326-362EA 181.28 USD
77326-362 77346-824 77362-668 77385-518
4-Methoxy-3,5-dimethylbenzamide ≥95%
4-Methoxy-3,5-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24038445
CAS Number: 1429903-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR