5-Bromo-1,3-dimethyl-2-propoxybenzene ≥95%
Supplier: AOBChem USA
|
Warning
|
37378-500MG
37378-1G
37378-5G
37378-25G
77326-358EA
199.82
USD
77326-358
77350-056
77362-664
77385-514
5-Bromo-1,3-dimethyl-2-propoxybenzene ≥95%
5-Bromo-1,3-dimethyl-2-propoxybenzene
Precaución: for Research Use Only
Formula:
C₁₁H₁₅BrO MW: 243.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18917815 CAS Number: 247080-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...