Ethyl 4-bromo-3-chloro-2-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

37376-500MG 37376-1G 37376-25G 37376-5G
77326-356EA 304.88 USD
77326-356 77350-054 77385-512 77374-274
Ethyl 4-bromo-3-chloro-2-fluorobenzoate ≥95%
Ethyl 4-bromo-3-chloro-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrClFO₂
MW: 281.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28733919
CAS Number: 1699416-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR