4-Bromo-3-chloro-N-cyclohexyl-2-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

37374-500MG 37374-1G 37374-5G
77326-352EA 317.24 USD
77326-352 77350-052 77374-270
4-Bromo-3-chloro-N-cyclohexyl-2-fluorobenzamide ≥97%
4-Bromo-3-chloro-N-cyclohexyl-2-fluorobenzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₄BrClFNO
MW: 334.612 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632208
CAS Number: 2404734-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR