4-Bromo-3-chloro-N-cyclopropyl-2-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

37372-500MG 37372-1G 37372-5G
77326-348EA 317.24 USD
77326-348 77350-048 77374-266
4-Bromo-3-chloro-N-cyclopropyl-2-fluorobenzamide ≥97%
4-Bromo-3-chloro-N-cyclopropyl-2-fluorobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₈BrClFNO
MW: 292.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632206
CAS Number: 2404733-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR