4-Bromo-3-chloro-2-fluoro-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

37371-500MG 37371-1G 37371-5G 37371-25G
77326-346EA 243.08 USD
77326-346 77350-046 77362-660 77385-510
4-Bromo-3-chloro-2-fluoro-N-methylbenzamide ≥97%
4-Bromo-3-chloro-2-fluoro-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₆BrClFNO
Storage Temperature: Ambient
MDL Number: MFCD32632205
CAS Number: 1860840-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR