4-Bromo-3-chloro-2-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

37370-500MG 37370-1G 37370-5G 37370-25G
77326-344EA 243.08 USD
77326-344 77350-044 77362-658 77385-508
4-Bromo-3-chloro-2-fluorobenzamide ≥97%
4-Bromo-3-chloro-2-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₄BrClFNO
MW: 252.468 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632204
CAS Number: 1863087-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR