(2-Fluoro-3-isopropoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

37355-1G 37355-5G 37355-25G
77341-716EA 101.97 USD
77341-716 77362-654 77374-264
(2-Fluoro-3-isopropoxyphenyl)(methyl)sulfane ≥95%
(2-Fluoro-3-isopropoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₃FOS
MW: 200.273 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632202
CAS Number: 2404733-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR