1,5-Dibromo-2-butoxy-3-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

37342-500MG 37342-1G 37342-5G 37342-25G
77326-338EA 87.55 USD
77326-338 77342-288 77362-650 77374-260
1,5-Dibromo-2-butoxy-3-fluorobenzene ≥95%
1,5-Dibromo-2-butoxy-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁Br₂FO
MW: 326 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632201
CAS Number: 2404733-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR