1,5-Dibromo-3-fluoro-2-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

37340-1G 37340-5G 37340-25G
77340-656EA 60.77 USD
77340-656 77362-648 77374-258
1,5-Dibromo-3-fluoro-2-propoxybenzene ≥97%
1,5-Dibromo-3-fluoro-2-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD32707021
CAS Number: 2413441-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR