3-Isopropoxy-5-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

37327-500MG 37327-1G 37327-5G
77326-336EA 317.24 USD
77326-336 77350-036 77374-254
3-Isopropoxy-5-(methylthio)pyridine ≥95%
3-Isopropoxy-5-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₃NOS
MW: 183.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632197
CAS Number: 2404734-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR