tert-Butyl 2,3-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

37320-500MG 37320-1G 37320-5G 37320-25G
77326-332EA 91.67 USD
77326-332 77342-430 77362-644 77374-252
tert-Butyl 2,3-difluorobenzoate ≥97%
tert-Butyl 2,3-difluorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₂
MW: 214.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18897595
CAS Number: 1339524-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR