3-Chloro-2-methoxy-4-(methoxymethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

37311-500MG 37311-1G 37311-5G
77335-798EA 366.68 USD
77335-798 77350-032 77374-250
3-Chloro-2-methoxy-4-(methoxymethyl)pyridine ≥95%
3-Chloro-2-methoxy-4-(methoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₂
MW: 187.624 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632195
CAS Number: 2404733-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR