2-Chloro-3-fluoro-4-(methoxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

37310-500MG 37310-1G 37310-5G 37310-25G
77326-330EA 128.75 USD
77326-330 77344-310 77362-642 77374-248
2-Chloro-3-fluoro-4-(methoxymethyl)pyridine ≥97%
2-Chloro-3-fluoro-4-(methoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₇ClFNO
MW: 175.588 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632194
CAS Number: 2404734-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR