3-(Difluoromethoxy)-4-fluoro-5-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

37298-500MG 37298-1G 37298-5G
77326-328EA 317.24 USD
77326-328 77350-030 77374-246
3-(Difluoromethoxy)-4-fluoro-5-methylbenzaldehyde ≥95%
3-(Difluoromethoxy)-4-fluoro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MW: 204.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632192
CAS Number: 2090318-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR