8-Iodo-6-methyl-[1,2,4]triazolo[4,3-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

37264-500MG 37264-1G 37264-5G
77326-322EA 341.96 USD
77326-322 77350-024 77374-242
8-Iodo-6-methyl-[1,2,4]triazolo[4,3-a]pyridine ≥97%
8-Iodo-6-methyl-[1,2,4]triazolo[4,3-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₆IN₃
MW: 259.047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632178
CAS Number: 2379322-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR