4-Bromo-2-chloro-3-fluorophenol ≥95%

Supplier: AOBChem USA

Warning

37262-1G 37262-5G 37262-25G
77341-478EA 89.61 USD
77341-478 77362-638 77374-240
4-Bromo-2-chloro-3-fluorophenol ≥95%
4-Bromo-2-chloro-3-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrClFO
MW: 225.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11849932
CAS Number: 1233026-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR