3-Chloro-2-fluoro-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37250-500MG 37250-1G 37250-5G
77340-016EA 713.79 USD
77340-016 77350-020 77374-236
3-Chloro-2-fluoro-5-methoxybenzoic acid ≥97%
3-Chloro-2-fluoro-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD20485132
CAS Number: 879541-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR