4-Chloro-1-(methoxymethyl)-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

37238-500MG 37238-1G 37238-5G 37238-25G
77326-318EA 119.48 USD
77326-318 77343-642 77362-636 77374-234
4-Chloro-1-(methoxymethyl)-2-(trifluoromethyl)benzene ≥97%
4-Chloro-1-(methoxymethyl)-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈ClF₃O
MW: 224.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22492225
CAS Number: 1824603-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR