(4-(Benzyloxy)-2,3-difluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

37237-500MG 37237-1G 37237-5G 37237-25G
77326-316EA 168.92 USD
77326-316 77346-322 77362-634 77385-494
(4-(Benzyloxy)-2,3-difluorophenyl)(methyl)sulfane ≥97%
(4-(Benzyloxy)-2,3-difluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂OS
Storage Temperature: Ambient
MDL Number: MFCD32632173
CAS Number: 2379321-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR