2,6-Dibromo-4-isopropylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37226-1G 37226-5G 37226-25G
77341-958EA 109.18 USD
77341-958 77362-630 77374-232
2,6-Dibromo-4-isopropylbenzaldehyde ≥97%
2,6-Dibromo-4-isopropylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀Br₂O
Storage Temperature: Ambient
MDL Number: MFCD32707016
CAS Number: 2484889-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR