2-Bromo-4-chloro-1-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

37211-1G 37211-5G 37211-25G
77340-654EA 60.77 USD
77340-654 77362-626 77374-228
2-Bromo-4-chloro-1-propoxybenzene ≥97%
2-Bromo-4-chloro-1-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
Storage Temperature: Ambient
MDL Number: MFCD16326677
CAS Number: 1225803-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR