1-Fluoro-2-isopropoxy-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

37189-1G 37189-5G 37189-25G
77341-104EA 71.07 USD
77341-104 77362-624 77374-226
1-Fluoro-2-isopropoxy-3-(trifluoromethyl)benzene ≥97%
1-Fluoro-2-isopropoxy-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₄O
MW: 222.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632155
CAS Number: 2379321-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR