Methyl 4-methoxy-3,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37175-500MG 37175-1G 37175-5G 37175-25G
77326-308EA 139.05 USD
77326-308 77344-660 77362-622 77385-488
Methyl 4-methoxy-3,5-dimethylbenzoate ≥97%
Methyl 4-methoxy-3,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025328
CAS Number: 70347-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR