2-Fluoro-4-methyl-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

37158-500MG 37158-1G 37158-5G 37158-25G
77326-302EA 222.48 USD
77326-302 77350-010 77362-616 77385-482
2-Fluoro-4-methyl-5-(methylthio)benzoic acid ≥97%
2-Fluoro-4-methyl-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD22490210
CAS Number: 958667-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR