(5-Bromo-2-fluoro-4-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

37157-1G 37157-5G 37157-25G
77341-456EA 87.55 USD
77341-456 77362-614 77374-222
(5-Bromo-2-fluoro-4-methylphenyl)(methyl)sulfane ≥95%
(5-Bromo-2-fluoro-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈BrFS
MW: 235.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632147
CAS Number: 2379322-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR