3,5-Dimethyl-1-(3-(trifluoromethyl)phenyl)-1H-pyrazole-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

37153-500MG 37153-1G 37153-5G 37153-25G
77326-300EA 193.64 USD
77326-300 77350-008 77362-612 77385-480
3,5-Dimethyl-1-(3-(trifluoromethyl)phenyl)-1H-pyrazole-4-carbaldehyde ≥97%
3,5-Dimethyl-1-(3-(trifluoromethyl)phenyl)-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₁F₃N₂O
MW: 268.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11927505
CAS Number: 1152866-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR