Ethyl 2-chloro-5-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

37127-1G 37127-5G 37127-25G
77340-990EA 66.95 USD
77340-990 77362-610 77374-220
Ethyl 2-chloro-5-methoxybenzoate ≥95%
Ethyl 2-chloro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
MW: 214.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396120
CAS Number: 1261792-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR