2-Ethoxy-1,4-difluoro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

37123-500MG 37123-1G 37123-25G 37123-5G
77326-296EA 292.52 USD
77326-296 77350-004 77385-476 77374-218
2-Ethoxy-1,4-difluoro-3-methoxybenzene ≥97%
2-Ethoxy-1,4-difluoro-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O₂
MW: 188.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632144
CAS Number: 2379322-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR