4-Bromo-2,5-difluoro-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

37121-500MG 37121-1G 37121-5G 37121-25G
77326-294EA 204.97 USD
77326-294 77350-002 77362-606 77385-474
4-Bromo-2,5-difluoro-4'-methyl-1,1'-biphenyl ≥97%
4-Bromo-2,5-difluoro-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₂
MW: 283.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564044
CAS Number: 204776-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR