2-Bromo-4'-chloro-4-fluoro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

37120-500MG 37120-1G 37120-5G
77326-292EA 341.96 USD
77326-292 77350-000 77374-216
2-Bromo-4'-chloro-4-fluoro-1,1'-biphenyl ≥95%
2-Bromo-4'-chloro-4-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇BrClF
Storage Temperature: Ambient
MDL Number: MFCD32632143
CAS Number: 2379321-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR