Methyl 3',5'-dimethyl-[1,1'-biphenyl]-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

37116-500MG 37116-1G 37116-5G 37116-25G
77326-288EA 119.48 USD
77326-288 77343-640 77362-602 77374-212
Methyl 3',5'-dimethyl-[1,1'-biphenyl]-4-carboxylate ≥97%
Methyl 3',5'-dimethyl-[1,1'-biphenyl]-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₂
MW: 240.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632140
CAS Number: 1075754-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR