2-(4-Bromo-3-fluorophenyl)-8-methylimidazo[1,2-a]pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
37081-500MG
37081-1G
37081-5G
77338-474EA
490.28
USD
77338-474
77349-994
77374-208
2-(4-Bromo-3-fluorophenyl)-8-methylimidazo[1,2-a]pyridine ≥97%
2-(4-Bromo-3-fluorophenyl)-8-methylimidazo[1,2-a]pyridine
Precaución: for Research Use Only
Formula:
C₁₄H₁₀BrFN₂ MW: 305.145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23650146 CAS Number: 1547439-45-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...