1-(4-Bromo-2-chlorophenyl)-1H-pyrrole ≥97%

Supplier: AOBChem USA

Warning

37072-500MG 37072-1G 37072-5G
77326-284EA 317.24 USD
77326-284 77349-990 77374-204
1-(4-Bromo-2-chlorophenyl)-1H-pyrrole ≥97%
1-(4-Bromo-2-chlorophenyl)-1H-pyrrole

Precaución: for Research Use Only

Formula: C₁₀H₇BrClN
MW: 256.526 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02665253
CAS Number: 383137-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR