2-(4-Chlorophenyl)-6-methylimidazo[1,2-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

37061-1G 37061-5G 37061-25G
77341-268EA 80.34 USD
77341-268 77362-596 77374-202
2-(4-Chlorophenyl)-6-methylimidazo[1,2-a]pyridine ≥97%
2-(4-Chlorophenyl)-6-methylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClN₂
MW: 242.704 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00444823
CAS Number: 103844-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR