2-(3-Bromophenyl)-6-methylimidazo[1,2-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

37057-500MG 37057-1G 37057-5G 37057-25G
77326-282EA 245.14 USD
77326-282 77349-988 77362-594 77385-468
2-(3-Bromophenyl)-6-methylimidazo[1,2-a]pyridine ≥97%
2-(3-Bromophenyl)-6-methylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrN₂
Storage Temperature: Ambient
MDL Number: MFCD01456302
CAS Number: 419557-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR