2-(3-Bromophenyl)-7-methylimidazo[1,2-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

37055-500MG 37055-1G 37055-5G 37055-25G
77326-280EA 245.14 USD
77326-280 77349-986 77362-592 77385-466
2-(3-Bromophenyl)-7-methylimidazo[1,2-a]pyridine ≥97%
2-(3-Bromophenyl)-7-methylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrN₂
MW: 287.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01456296
CAS Number: 419557-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR