2-(Benzyloxy)-4-chloro-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37044-500MG 37044-1G 37044-5G
77337-062EA 407.88 USD
77337-062 77349-982 77374-200
2-(Benzyloxy)-4-chloro-3-fluorobenzaldehyde ≥97%
2-(Benzyloxy)-4-chloro-3-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClFO₂
MW: 264.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902598
CAS Number: 2586125-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR