3-Fluoro-2-methoxy-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37034-500MG 37034-1G 37034-5G 37034-25G
77326-272EA 247.2 USD
77326-272 77349-980 77362-584 77385-462
3-Fluoro-2-methoxy-4-methylbenzoic acid ≥97%
3-Fluoro-2-methoxy-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD23712093
CAS Number: 1427416-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR